Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Human IgG1 Fc (C103S), Avitag™,His Tag (IG1-H82E9) is expressed from human 293 cells (HEK293). It contains AA Pro 100 - Lys 330 (Accession # P01857-1 (C103S)). 1 * 200 µG
Catalog # ACRBIG1-H82E9200UG
Supplier:  ACRO Biosystems
Biotinylated Human IgG1 Fc (C103S), Avitag™,His Tag (IG1-H82E9) is expressed from human 293 cells (HEK293). It contains AA Pro 100 - Lys 330 (Accession # P01857-1 (C103S)). 1 * 200 µG
Catalog # ACRBIG1-H82E9200UG
Supplier:  ACRO Biosystems
Frequently Bought Together