Order Entry
Slovakia
ContactUsLinkComponent
Mouse IL-17C, His Tag (NON-H5219) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Cys 232 (Accession # Q13253-1) . 1 * 20 µG
Katalógové číslo ACRBNON-H5219-20UG
Supplier:  ACRO Biosystems
Mouse IL-17C, His Tag (NON-H5219) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Cys 232 (Accession # Q13253-1) . 1 * 20 µG
Katalógové číslo ACRBNON-H5219-20UG
Supplier:  ACRO Biosystems
Frequently Bought Together