Digital Solutions
Applications
Support
About
Slovakia
ContactUsLinkComponent
Rat B7-H2, Fc Tag (B72-R5259) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Lys 261 (Accession # XP_006256322. 1). 1 * 100 µG
ACRBB72-R5259100UG
:
ACRO Biosystems
Rat B7-H2, Fc Tag (B72-R5259) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Lys 261 (Accession # XP_006256322. 1). 1 * 100 µG
ACRBB72-R5259100UG
:
ACRO Biosystems
Frequently Bought Together
VWR®, Cryoboxes, Cardboard, 136x136 mm
: VWR Collection
Cryogenic vials, Cryovial®
: SIMPORT SCIENTIFIC
VWR®, Roller Bottles
: VWR Collection
Push-in stoppers for tubes
: SIMPORT SCIENTIFIC
Individual PCR tubes with attached caps
: BRAND
VWR®, Blotting Pads, Grade 707
: VWR Collection
Screw caps for SuperClear® microtubes
: Labcon
Centrifuge tubes with flat caps, MetalFree®
: Labcon
Test tubes
: DELTALAB
Microplate 1536 well polystyrene white F-bottom HI-base non-binding non-sterile (15 pieces/bag) 1 * 60 items
: Greiner Bio-One