Order Entry
Slovakia
ContactUsLinkComponent
Rat B7-1, His Tag (B71-R52H9) is expressed from human 293 cells (HEK293). It contains AA Ile 39 - Gln 248 (Accession # Q62680-1). 1 * 100 µG
Catalog # ACRBB71-R52H9100UG
Supplier:  ACRO Biosystems
Rat B7-1, His Tag (B71-R52H9) is expressed from human 293 cells (HEK293). It contains AA Ile 39 - Gln 248 (Accession # Q62680-1). 1 * 100 µG
Catalog # ACRBB71-R52H9100UG
Supplier:  ACRO Biosystems
Frequently Bought Together