Zadaní objednávky
Slovakia
ContactUsLinkComponent
Mouse Plasma Kallikrein/KLKB1, His Tag (KL1-M52H9) is expressed from human 293 cells (HEK293). It contains AA Gly 20 - Ala 638 (Acce ssion # P26262-1). 1 * 100 µG
Catalog # ACRBKL1-M52H9100UG
Supplier:  ACRO Biosystems
Mouse Plasma Kallikrein/KLKB1, His Tag (KL1-M52H9) is expressed from human 293 cells (HEK293). It contains AA Gly 20 - Ala 638 (Acce ssion # P26262-1). 1 * 100 µG
Catalog # ACRBKL1-M52H9100UG
Supplier:  ACRO Biosystems
Frequently Bought Together