GEMPURE SELECT (USP) WFI WATER 1L SAMPLE 1 * 1 L
Catalog # GEMB900-001-001
Supplier: GEMINI BIO-PRODUCTS
GEMPURE SELECT (USP) WFI WATER 1L SAMPLE 1 * 1 L
Catalog # GEMB900-001-001
Supplier: GEMINI BIO-PRODUCTS
Frequently Bought Together
VWR®, Spreaders, T-shaped
Supplier: VWR Collection
Pasteur pipettes, disposable
Supplier: BRAND
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR® Disposable Microcentrifuge Tubes
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Freestanding, with Flat Screw Caps
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection