Order Entry
Slovakia
ContactUsLinkComponent
Hydrochloric Acid 1M (1N) CRM, ISO 17034 and ISO 17025,,,, Conservation 24 months. ,,,, Store under normal laboratory conditions, at temperatures between 15°C to 25°C,,,, 1 * 1 L
Katalógové číslo CMPLZ13411027.1L
Hydrochloric Acid 1M (1N) CRM, ISO 17034 and ISO 17025,,,, Conservation 24 months. ,,,, Store under normal laboratory conditions, at temperatures between 15°C to 25°C,,,, 1 * 1 L
Katalógové číslo CMPLZ13411027.1L
Frequently Bought Together