Assembled eccentric group with rev counter 1 * 1 items
Catalog # VELP40001839
Supplier: VELP SCIENTIFIC
Assembled eccentric group with rev counter 1 * 1 items
Catalog # VELP40001839
Supplier: VELP SCIENTIFIC
Frequently Bought Together
Schutzfilter 5 ml für Eppendorf Research Plus 1 * 50 ST
Supplier: EPPENDORF
Fixing cover device 1 * 1 items
Supplier: VWR Collection
Lithium Polymer Rechargable Battery
Supplier: EPPENDORF
Sacrifical body 1 * 1 items
Supplier: VWR Collection
Accessories for electronic dispensers, Multipette E3/E3x
Supplier: EPPENDORF
Overpressur plug 1 * 1 items
Supplier: VWR Collection
Accessories for PIPETBOY acu / pro
Supplier: INTEGRA BIOSCIENCE
Accessories for HandyStep® electronic
Supplier: BRAND
VWR®, Accessories for Mechanical Pipettes
Supplier: VWR Collection
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
Pressure regulator 1 * 1 items
Supplier: VWR Collection
BIOTECTOR BASE REAGENT 1 * 20 L
Supplier: HACH
AIR FILTERDIA.50x1,8 NPT LABOKLAV 1 * 1 items
Supplier: SHP Steriltechnik
Accessories for Diaphragm Pumps
Supplier: KNF
Suction tube 6x300ml kpl. 1 * 1 items
Supplier: BUCHI
Vacuum seal PTFE / FKM
Supplier: Heidolph Instruments GmbH & Co.KG
BIOTECTOR ACID REAGENT WITH CATALYST 1 * 20 L
Supplier: HACH
Accessories
Supplier: BRAND