Order Entry
Slovakia
ContactUsLinkComponent
IR-CO2 sensor ES, Infrared sensor for measuring carbon ,dioxide concentrations in the ambient air.For use in the Dräger X-am 5600 and, Dräger X-am 8000. Measuring range: 0-5 Vol% CO2 1 * 1 items
Catalog # DRAE6851882
Supplier:  Dräger
IR-CO2 sensor ES, Infrared sensor for measuring carbon ,dioxide concentrations in the ambient air.For use in the Dräger X-am 5600 and, Dräger X-am 8000. Measuring range: 0-5 Vol% CO2 1 * 1 items
Catalog # DRAE6851882
Supplier:  Dräger
Frequently Bought Together