CURZERENE, PHYPROOF(R) REFERENCE SUBSTANCE 1 * 10 mg
Catalog # SIALPHL84768-10MG
Supplier: Merck
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solution, pH 9 (at 20 °C), Borate free
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
UV/Visible standards, Supelco®
Supplier: Merck
ICP multi-element standards, CertiPUR®, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Supelco® TraceCERT® Organic Reference Standard, Tebutam
Supplier: Merck
pH Standard Solutions, IUPAC, Certified
Supplier: HACH
Reagents for chloride determination using Lovibond® MD photometers
Supplier: TINTOMETER