Order Entry
Slovakia
ContactUsLinkComponent
Sanitary End Cap, PP; 1.5" Sanitary Tri-Clamp
Sanitary End Cap, PP; 1.5" Sanitary Tri-Clamp
Catalog # MFLX06320-81
undefined
Sanitary End Cap, PP; 1.5" Sanitary Tri-Clamp
Catalog # MFLX06320-81
About this item
Create a secure connection in your sanitary fitting system.
  • Constructed of durable polypropylene (PP)
Close off your sanitary fluid line with these convenient end caps.
  • Connection Type:
    Sanitary Cap
  • Flow pattern:
    Straight
  • Sanitary clamp size (Port 1):
    1-1/2"
  • Description:
    Sanitary End Cap, PP; 1-1/2" Sanitary Tri-Clamp
  • Environmentally Preferable:
  • Warranty:
    6 Months
  • Pk:
    1
  • Fitting material:
    Polypropylene
  • Sanitary gasket size:
    1-1/2"
Frequently Bought Together