Order Entry
Slovakia
ContactUsLinkComponent
Reagecon 0.5 mg/L C from USP Sucrose Total Organic Carbon (TOC) Standard for Analytik Jena, Lachat, OI Analytical, Teledyne Tekmar and Shimadzu TOC-L/TOC- V/TOC-Vcsh/TOC-4200 Analysers 1 * 250 mL
Catalog # REGEISTOC1126
Supplier:  REAGECON
Reagecon 0.5 mg/L C from USP Sucrose Total Organic Carbon (TOC) Standard for Analytik Jena, Lachat, OI Analytical, Teledyne Tekmar and Shimadzu TOC-L/TOC- V/TOC-Vcsh/TOC-4200 Analysers 1 * 250 mL
Catalog # REGEISTOC1126
Supplier:  REAGECON
Frequently Bought Together