Zadaní objednávky
Slovakia
ContactUsLinkComponent
Software conforming to FDA AtmoCONTROL. Meets the requirements for the use of electronically stored data sets and electronic signatu res as laid down in Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA). Respective IQ/OQ document 1 * 1 items
Catalog # MEMMFDAQ1UN750MPLS
Supplier:  MEMMERT
Software conforming to FDA AtmoCONTROL. Meets the requirements for the use of electronically stored data sets and electronic signatu res as laid down in Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA). Respective IQ/OQ document 1 * 1 items
Catalog # MEMMFDAQ1UN750MPLS
Supplier:  MEMMERT
Frequently Bought Together