Order Entry
Slovakia
ContactUsLinkComponent
Human Tissue Factor, Fc Tag (TF3-H5253) is expressed from human 293 cells (HEK293). It contains AA Ser 33 - Glu 251 (Accession # P13 726-1). 1 * 20 µG
Catalog # ACRBTF3-H5253-20UG
Supplier:  ACRO Biosystems
Human Tissue Factor, Fc Tag (TF3-H5253) is expressed from human 293 cells (HEK293). It contains AA Ser 33 - Glu 251 (Accession # P13 726-1). 1 * 20 µG
Catalog # ACRBTF3-H5253-20UG
Supplier:  ACRO Biosystems
Frequently Bought Together