Order Entry
Slovakia
ContactUsLinkComponent
Human CCL5, Fc Tag (CC5-H5251) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Ser 91 (Accession # P13501-1). 1 * 50 µG
Catalog # ACRBCC5-H5251-50UG
Dodávateľ:  ACRO Biosystems
Human CCL5, Fc Tag (CC5-H5251) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Ser 91 (Accession # P13501-1). 1 * 50 µG
Catalog # ACRBCC5-H5251-50UG
Dodávateľ:  ACRO Biosystems
Frequently Bought Together