Order Entry
Slovakia
ContactUsLinkComponent
Human PD-1, His tag (Nanoparticle) (PD1-H52H2) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accessio n # Q15116-1). 1 * 500 µG
Catalog # ACRBPD1-H52H2500UG
Supplier:  ACRO Biosystems
Human PD-1, His tag (Nanoparticle) (PD1-H52H2) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accessio n # Q15116-1). 1 * 500 µG
Catalog # ACRBPD1-H52H2500UG
Supplier:  ACRO Biosystems
Frequently Bought Together