Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Mouse TGF-Beta 1, Avitag (TG1-M8218) is expressed from human 293 cells (HEK293). It contains AA Ala 279 - Ser 390 (Acce ssion # P04202-1). 1 * 100 µG
Catalog # ACRBTG1-M8218100UG
Supplier:  ACRO Biosystems
Biotinylated Mouse TGF-Beta 1, Avitag (TG1-M8218) is expressed from human 293 cells (HEK293). It contains AA Ala 279 - Ser 390 (Acce ssion # P04202-1). 1 * 100 µG
Catalog # ACRBTG1-M8218100UG
Supplier:  ACRO Biosystems
Frequently Bought Together