Zadaní objednávky
Slovakia
ContactUsLinkComponent
Biotinylated Human CD40 Ligand, Avitag,His,Flag Tag (CDL-H82Db) is expressed from human 293 cells (HEK293). It contains AA Gly 116 - Leu 261 (Accession # P29965-1). 1 * 25 µG
Catalog # ACRBCDL-H82DB-25UG
Supplier:  ACRO Biosystems
Biotinylated Human CD40 Ligand, Avitag,His,Flag Tag (CDL-H82Db) is expressed from human 293 cells (HEK293). It contains AA Gly 116 - Leu 261 (Accession # P29965-1). 1 * 25 µG
Catalog # ACRBCDL-H82DB-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together