Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Mouse Serum Albumin, His,Avitag (MSA-M82E4) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Ala 608 (Accession # P07724-1). 1 * 25 µG
  ACRBMSA-M82E4-25UG
Biotinylated Mouse Serum Albumin, His,Avitag (MSA-M82E4) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Ala 608 (Accession # P07724-1). 1 * 25 µG
  ACRBMSA-M82E4-25UG
Frequently Bought Together