Order Entry
Slovakia
ContactUsLinkComponent
Complete Denaturing Gradient Gel Electrophoresis System, 20x20cm include: temperature control unit, cam castingbase, glass plates with 1mm bonded spacers,2x 24-sample combs and gradient mixer -240 VAC version 1 * 1 KIT
Catalog # CLEVVS20WAVE-DGGE
Supplier:  CLEAVER SCIENTIFIC
Complete Denaturing Gradient Gel Electrophoresis System, 20x20cm include: temperature control unit, cam castingbase, glass plates with 1mm bonded spacers,2x 24-sample combs and gradient mixer -240 VAC version 1 * 1 KIT
Catalog # CLEVVS20WAVE-DGGE
Supplier:  CLEAVER SCIENTIFIC
Frequently Bought Together