Order Entry
Slovakia
ContactUsLinkComponent
Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1 is produced from a hybridoma resulting from fusion of SP20 myeloma and B-lymphocytes obtained from a mouse immunized with FMC63 scFv. 1 * 250 Tests
Catalog # ACRBFM3-Y45-250TST
Supplier:  ACRO Biosystems
Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1 is produced from a hybridoma resulting from fusion of SP20 myeloma and B-lymphocytes obtained from a mouse immunized with FMC63 scFv. 1 * 250 Tests
Catalog # ACRBFM3-Y45-250TST
Supplier:  ACRO Biosystems
Frequently Bought Together