Order Entry
Slovakia
ContactUsLinkComponent
Anti-BCMA Neutralizing Antibody, Mouse IgG1 BCA-M43 recognizes the Human BCMA Protein extracellular and can inhibit the interaction between BCMA and BAFFAPRIL. It is expressed from human 293 cells HEK293. 1 * 100 µG
Catalog # ACRBBCA-M43-100UG
Supplier:  ACRO Biosystems
Anti-BCMA Neutralizing Antibody, Mouse IgG1 BCA-M43 recognizes the Human BCMA Protein extracellular and can inhibit the interaction between BCMA and BAFFAPRIL. It is expressed from human 293 cells HEK293. 1 * 100 µG
Catalog # ACRBBCA-M43-100UG
Supplier:  ACRO Biosystems
Frequently Bought Together