Order Entry
Slovakia
ContactUsLinkComponent
Cynomolgus TSLP, His Tag TSP-C52H8 is expressed from human 293 cells HEK293. It contains AA Tyr 29 - Gln 159 Accession A0A2K5TXV0-1 . 1 * 100 µG
Catalog # ACRBTSP-C52H8100UG
Supplier:  ACRO Biosystems
Cynomolgus TSLP, His Tag TSP-C52H8 is expressed from human 293 cells HEK293. It contains AA Tyr 29 - Gln 159 Accession A0A2K5TXV0-1 . 1 * 100 µG
Catalog # ACRBTSP-C52H8100UG
Supplier:  ACRO Biosystems
Frequently Bought Together