Order Entry
Slovakia
ContactUsLinkComponent
Rhesus macaque CD32a, His Tag SPR  BLI verified CDA-R52H4 is expressed from human 293 cells HEK293. It contains AA Gln 34 - Ile 217 Accession NP_001244229.1. 1 * 1 mg
Catalog # ACRBCDA-R52H4-1MG
Supplier:  ACRO Biosystems
Rhesus macaque CD32a, His Tag SPR  BLI verified CDA-R52H4 is expressed from human 293 cells HEK293. It contains AA Gln 34 - Ile 217 Accession NP_001244229.1. 1 * 1 mg
Catalog # ACRBCDA-R52H4-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together