Zadaní objednávky
Slovakia
ContactUsLinkComponent
Biotinylated SARS-CoV-2 S protein RBD, His,Avitag SPD-C82E9 is expressed from human 293 cells HEK293. It contains AA Arg 319 - Phe 5 41 Accession QHD43416.1. 1 * 25 µG
Catalog # ACRBSPD-C82E9-25UG
Dodávateľ:  ACRO Biosystems
Biotinylated SARS-CoV-2 S protein RBD, His,Avitag SPD-C82E9 is expressed from human 293 cells HEK293. It contains AA Arg 319 - Phe 5 41 Accession QHD43416.1. 1 * 25 µG
Catalog # ACRBSPD-C82E9-25UG
Dodávateľ:  ACRO Biosystems
Frequently Bought Together