Order Entry
Slovakia
ContactUsLinkComponent
Human ACE2, His Tag AC2-H52H8 is expressed from human 293 cells HEK293. It contains AA Gln 18 - Ser 740 Accession Q9BYF1-1. 1 * 50 µG
Catalog # ACRBAC2-H52H8-50UG
Supplier:  ACRO Biosystems
Human ACE2, His Tag AC2-H52H8 is expressed from human 293 cells HEK293. It contains AA Gln 18 - Ser 740 Accession Q9BYF1-1. 1 * 50 µG
Catalog # ACRBAC2-H52H8-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together