Order Entry
Slovakia
ContactUsLinkComponent
Human CD40 Ligand, Fc Tag active trimer MALS verified CDL-H5269 is expressed from human 293 cells HEK293. It contains AA Gly 116 - L eu 261 Accession P29965-1 trimer Design. 1 * 100 µG
Katalógové číslo ACRBCDL-H5269100UG
Supplier:  ACRO Biosystems
Human CD40 Ligand, Fc Tag active trimer MALS verified CDL-H5269 is expressed from human 293 cells HEK293. It contains AA Gly 116 - L eu 261 Accession P29965-1 trimer Design. 1 * 100 µG
Katalógové číslo ACRBCDL-H5269100UG
Supplier:  ACRO Biosystems
Frequently Bought Together