Order Entry
Slovakia
ContactUsLinkComponent
Mouse ACE2, His Tag AC2-M5248 is expressed from human 293 cells HEK293. It contains AA Gln 18 - Thr 740 Accession Q8R0I0-1. 1 * 50 µG
Catalog # ACRBAC2-M5248-50UG
Supplier:  ACRO Biosystems
Mouse ACE2, His Tag AC2-M5248 is expressed from human 293 cells HEK293. It contains AA Gln 18 - Thr 740 Accession Q8R0I0-1. 1 * 50 µG
Catalog # ACRBAC2-M5248-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together