Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Mouse CD28, His,Avitag CD8-M82E3 is expressed from human 293 cells HEK293. It contains AA Asn 20 - Lys 149 Accession P 31041-1. 1 * 25 µG
Catalog # ACRBCD8-M82E3-25UG
Supplier:  ACRO Biosystems
Biotinylated Mouse CD28, His,Avitag CD8-M82E3 is expressed from human 293 cells HEK293. It contains AA Asn 20 - Lys 149 Accession P 31041-1. 1 * 25 µG
Catalog # ACRBCD8-M82E3-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together