Order Entry
Slovakia
ContactUsLinkComponent
SARS-CoV-2 S protein RBD W436R, His Tag SPD-S52H7 is expressed from human 293 cells HEK293. It contains AA Arg 319 - Phe 541 Accessi on QHD43416.1 W436R. 1 * 1 mg
Catalog # ACRBSPD-S52H7-1MG
Dodávateľ:  ACRO Biosystems
SARS-CoV-2 S protein RBD W436R, His Tag SPD-S52H7 is expressed from human 293 cells HEK293. It contains AA Arg 319 - Phe 541 Accessi on QHD43416.1 W436R. 1 * 1 mg
Catalog # ACRBSPD-S52H7-1MG
Dodávateľ:  ACRO Biosystems
Frequently Bought Together