Order Entry
Slovakia
ContactUsLinkComponent
Rhesus macaque CTGF, His Tag GTF-R52H4 is expressed from human 293 cells HEK293. It contains AA Gln 27 - Ala 349 Accession H9FQD5-1 . 1 * 100 µG
Catalog # ACRBGTF-R52H4100UG
Supplier:  ACRO Biosystems
Rhesus macaque CTGF, His Tag GTF-R52H4 is expressed from human 293 cells HEK293. It contains AA Gln 27 - Ala 349 Accession H9FQD5-1 . 1 * 100 µG
Catalog # ACRBGTF-R52H4100UG
Supplier:  ACRO Biosystems
Frequently Bought Together