Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Human CTLA-4, His,Avitag CT4-H82E3 is expressed from human 293 cells HEK293. It contains AA Ala 37 - Phe 162 Accession Q6GR94-1. 1 * 200 µG
Catalog # ACRBCT4-H82E3200UG
Supplier:  ACRO Biosystems
Biotinylated Human CTLA-4, His,Avitag CT4-H82E3 is expressed from human 293 cells HEK293. It contains AA Ala 37 - Phe 162 Accession Q6GR94-1. 1 * 200 µG
Catalog # ACRBCT4-H82E3200UG
Supplier:  ACRO Biosystems
Frequently Bought Together