Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Human TNFSF11, His,Avitag, active trimer RAL-H82Q5 is expressed from human 293 cells HEK293. It contains AA Gly 64 - As p 245 Accession AAC51762.1 trimer Design. 1 * 200 µG
Catalog # ACRBRAL-H82Q5200UG
Supplier:  ACRO Biosystems
Biotinylated Human TNFSF11, His,Avitag, active trimer RAL-H82Q5 is expressed from human 293 cells HEK293. It contains AA Gly 64 - As p 245 Accession AAC51762.1 trimer Design. 1 * 200 µG
Catalog # ACRBRAL-H82Q5200UG
Supplier:  ACRO Biosystems
Frequently Bought Together