Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Human IL-5, His,Avitag IL5-H82Q5 is expressed from human 293 cells HEK293. It contains AA Ile 20 - Ser 134 Accession P 05113-1. 1 * 25 µG
Catalog # ACRBIL5-H82Q5-25UG
Supplier:  ACRO Biosystems
Biotinylated Human IL-5, His,Avitag IL5-H82Q5 is expressed from human 293 cells HEK293. It contains AA Ile 20 - Ser 134 Accession P 05113-1. 1 * 25 µG
Catalog # ACRBIL5-H82Q5-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together