Order Entry
Slovakia
ContactUsLinkComponent
Cynomolgus FOLR2, His Tag FO2-C52H8 is expressed from human 293 cells HEK293. It contains AA Thr 34 - His 245 Accession A0A2K5U027- 1. 1 * 100 µG
Catalog # ACRBFO2-C52H8100UG
Supplier:  ACRO Biosystems
Cynomolgus FOLR2, His Tag FO2-C52H8 is expressed from human 293 cells HEK293. It contains AA Thr 34 - His 245 Accession A0A2K5U027- 1. 1 * 100 µG
Catalog # ACRBFO2-C52H8100UG
Supplier:  ACRO Biosystems
Frequently Bought Together