Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Human TNFR2, His,Avitag™ TN2-H82E3 is expressed from human 293 cells HEK293. It contains AA Leu 23 - Asp 257 Accession P20333-1. 1 * 25 µG
Catalog # ACRBTN2-H82E3-25UG
Supplier:  ACRO Biosystems
Biotinylated Human TNFR2, His,Avitag™ TN2-H82E3 is expressed from human 293 cells HEK293. It contains AA Leu 23 - Asp 257 Accession P20333-1. 1 * 25 µG
Catalog # ACRBTN2-H82E3-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together