Order Entry
Slovakia
ContactUsLinkComponent
Human RNF43, Fc Tag RN3-H5256 is expressed from human 293 cells HEK293. It contains AA Gln 44 - Tyr 197 Accession Q68DV7-1. 1 * 100 µG
Catalog # ACRBRN3-H5256100UG
Supplier:  ACRO Biosystems
Human RNF43, Fc Tag RN3-H5256 is expressed from human 293 cells HEK293. It contains AA Gln 44 - Tyr 197 Accession Q68DV7-1. 1 * 100 µG
Catalog # ACRBRN3-H5256100UG
Supplier:  ACRO Biosystems
Frequently Bought Together