Order Entry
Slovakia
ContactUsLinkComponent
SARS-CoV-2 S protein RBD L452R, His Tag SPD-C52He is expressed from human 293 cells HEK293. It contains AA Arg 319 - Lys 537L452R Ac cession QHD43416.1L452R. 1 * 1 mg
Catalog # ACRBSPD-C52HE-1MG
Supplier:  ACRO Biosystems
SARS-CoV-2 S protein RBD L452R, His Tag SPD-C52He is expressed from human 293 cells HEK293. It contains AA Arg 319 - Lys 537L452R Ac cession QHD43416.1L452R. 1 * 1 mg
Catalog # ACRBSPD-C52HE-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together