Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Human CLEC12A, Fc,Avitag CLA-H82F7 is expressed from human 293 cells HEK293. It contains AA His 65 - Ala 265 Accession Q5QGZ9-2. 1 * 200 µG
Catalog # ACRBCLA-H82F7200UG
Supplier:  ACRO Biosystems
Biotinylated Human CLEC12A, Fc,Avitag CLA-H82F7 is expressed from human 293 cells HEK293. It contains AA His 65 - Ala 265 Accession Q5QGZ9-2. 1 * 200 µG
Catalog # ACRBCLA-H82F7200UG
Supplier:  ACRO Biosystems
Frequently Bought Together