Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated SARS-CoV-2 S protein RBD, His,Avitag SPD-C82E8 is expressed from human 293 cells HEK293. It contains AA Arg 319 - Phe 5 41 Accession QHD43416.1. 1 * 1 mg
Catalog # ACRBSPD-C82E8-1MG
Supplier:  ACRO Biosystems
Biotinylated SARS-CoV-2 S protein RBD, His,Avitag SPD-C82E8 is expressed from human 293 cells HEK293. It contains AA Arg 319 - Phe 5 41 Accession QHD43416.1. 1 * 1 mg
Catalog # ACRBSPD-C82E8-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together