Order Entry
Slovakia
ContactUsLinkComponent
Nucleocapsid N protein is the most abundant protein found in coronavirus. CoV N protein is a highly immunogenic phosphoprotein impor tant for viral genome replication and modulation of cell signaling pathways. It was first identified by a research te 1 * 2 mg
  ACRBMBS-K017-2MG
Nucleocapsid N protein is the most abundant protein found in coronavirus. CoV N protein is a highly immunogenic phosphoprotein impor tant for viral genome replication and modulation of cell signaling pathways. It was first identified by a research te 1 * 2 mg
  ACRBMBS-K017-2MG
Frequently Bought Together