Order Entry
Slovakia
ContactUsLinkComponent
FITC-Labeled Human EpCAM, Fc Tag EPM-HF255 is expressed from human 293 cells HEK293. It contains AA Gln 24 - Lys 265 Accession AAH1 4785.1. 1 * 200 µG
Catalog # ACRBEPM-HF255200UG
Supplier:  ACRO Biosystems
FITC-Labeled Human EpCAM, Fc Tag EPM-HF255 is expressed from human 293 cells HEK293. It contains AA Gln 24 - Lys 265 Accession AAH1 4785.1. 1 * 200 µG
Catalog # ACRBEPM-HF255200UG
Supplier:  ACRO Biosystems
Frequently Bought Together