Order Entry
Slovakia
ContactUsLinkComponent
Human CD16a V176, HSA,His Tag CDA-H52S1 is expressed from human 293 cells HEK293. It contains AA Gly 17 - Gln 208 Accession P08637- 1 F176V. 1 * 1 mg
Catalog # ACRBCDA-H52S1-1MG
Supplier:  ACRO Biosystems
Human CD16a V176, HSA,His Tag CDA-H52S1 is expressed from human 293 cells HEK293. It contains AA Gly 17 - Gln 208 Accession P08637- 1 F176V. 1 * 1 mg
Catalog # ACRBCDA-H52S1-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together