Order Entry
Slovakia
ContactUsLinkComponent
Human LIV-1, His Tag LV1-H5223 is expressed from human 293 cells HEK293. It contains AA Phe 29 - Trp 325 Accession Q13433-1. 1 * 100 µG
Catalog # ACRBLV1-H5223100UG
Supplier:  ACRO Biosystems
Human LIV-1, His Tag LV1-H5223 is expressed from human 293 cells HEK293. It contains AA Phe 29 - Trp 325 Accession Q13433-1. 1 * 100 µG
Catalog # ACRBLV1-H5223100UG
Supplier:  ACRO Biosystems
Frequently Bought Together