Order Entry
Slovakia
ContactUsLinkComponent
SARS-CoV-2 S1 protein NTD A262S, His Tag S1D-C52H5 is expressed from human 293 cells HEK293. It contains AA Ser 13 - Leu 303 Access ion QHD43416.1 A262S. 1 * 1 mg
Catalog # ACRBS1D-C52H5-1MG
Supplier:  ACRO Biosystems
SARS-CoV-2 S1 protein NTD A262S, His Tag S1D-C52H5 is expressed from human 293 cells HEK293. It contains AA Ser 13 - Leu 303 Access ion QHD43416.1 A262S. 1 * 1 mg
Catalog # ACRBS1D-C52H5-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together