Order Entry
Slovakia
ContactUsLinkComponent
Human IL-1 Rrp2 C154S, C262S, His Tag IL2-H52H4 is expressed from human 293 cells HEK293. It contains AA Asp 20 - Arg 335 Accession Q9HB29-1 C154S, C262S. 1 * 100 µG
Catalog # ACRBIL2-H52H4100UG
Supplier:  ACRO Biosystems
Human IL-1 Rrp2 C154S, C262S, His Tag IL2-H52H4 is expressed from human 293 cells HEK293. It contains AA Asp 20 - Arg 335 Accession Q9HB29-1 C154S, C262S. 1 * 100 µG
Catalog # ACRBIL2-H52H4100UG
Supplier:  ACRO Biosystems
Frequently Bought Together