Order Entry
Slovakia
ContactUsLinkComponent
Rabbit IgG Fc, Tag Free IGG-R5203 is expressed from human 293 cells HEK293. It contains AA Cys 105 - Lys 323 Accession P01870-1. 1 * 1 mg
Catalog # ACRBIGG-R5203-1MG
Supplier:  ACRO Biosystems
Rabbit IgG Fc, Tag Free IGG-R5203 is expressed from human 293 cells HEK293. It contains AA Cys 105 - Lys 323 Accession P01870-1. 1 * 1 mg
Catalog # ACRBIGG-R5203-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together