Order Entry
Slovakia
ContactUsLinkComponent
Biotinylated Human IL-8, His,Avitag IL8-H82H3 is expressed from human 293 cells HEK293. It contains AA Ser 28 - Ser 99 Accession P1 0145-1. 1 * 25 µG
Catalog # ACRBIL8-H82H3-25UG
Supplier:  ACRO Biosystems
Biotinylated Human IL-8, His,Avitag IL8-H82H3 is expressed from human 293 cells HEK293. It contains AA Ser 28 - Ser 99 Accession P1 0145-1. 1 * 25 µG
Catalog # ACRBIL8-H82H3-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together