Order Entry
Slovakia
ContactUsLinkComponent
PE-Labeled Human Siglec-3, His Avitag CD3-HP2E3 is expressed from human 293 cells HEK293. It contains AA Asp 18 - His 259 Accession AAH28152.1. 1 * 200 Tests
Catalog # ACRBCD3HP2E3200TST
Supplier:  ACRO Biosystems
PE-Labeled Human Siglec-3, His Avitag CD3-HP2E3 is expressed from human 293 cells HEK293. It contains AA Asp 18 - His 259 Accession AAH28152.1. 1 * 200 Tests
Catalog # ACRBCD3HP2E3200TST
Supplier:  ACRO Biosystems
Frequently Bought Together