Order Entry
Slovakia
ContactUsLinkComponent
Human Netrin receptor DCC, Fc Tag NEC-H5254 is expressed from human 293 cells HEK293. It contains AA Phe 32 - Asn 1097 Accession P4 3146-1. 1 * 100 µG
Catalog # ACRBNEC-H5254100UG
Supplier:  ACRO Biosystems
Human Netrin receptor DCC, Fc Tag NEC-H5254 is expressed from human 293 cells HEK293. It contains AA Phe 32 - Asn 1097 Accession P4 3146-1. 1 * 100 µG
Catalog # ACRBNEC-H5254100UG
Supplier:  ACRO Biosystems
Frequently Bought Together